(L)-4,4,4,-Trifluoro-α-homoalanine, N-FMOC protected 98%
Supplier: Ambeed
|
Warning
|
A387334-1G
A387334-250MG
A387334-5G
A387334-100MG
76817-740EA
420
USD
76817-740
76817-742
77551-148
77884-024
(L)-4,4,4,-Trifluoro-α-homoalanine, N-FMOC protected 98%
(L)-4,4,4,-Trifluoro-α-homoalanine, N-FMOC protected
Precaución: for Research Use Only
Formula:
C₁₉H₁₆F₃NO₄ |
MDL Number:
MFCD08532485 CAS Number: 181128-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...