2-(Methoxymethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

A387053-10G A387053-1G A387053-25G A387053-5G
76817-736EA 153.84 USD
76817-736 76817-738 77572-154 77572-156
2-(Methoxymethoxy)phenylboronic acid 98%
2-(Methoxymethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD10566597
CAS Number: 115377-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR