Ethyl 5-(4-Aminophenyl)oxazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A386671-100MG A386671-1G A386671-250MG
76817-724EA 49 USD
76817-724 76817-726 76817-728
Ethyl 5-(4-Aminophenyl)oxazole-4-carboxylate 95%
Ethyl 5-(4-Aminophenyl)oxazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃
MDL Number: MFCD22200982
CAS Number: 950603-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR