(3-Methylbutyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Isopentylboronic acid

A386271-1G A386271-25G A386271-5G
76817-712EA 12 USD
76817-712 76817-714 76817-716
(3-Methylbutyl)boronic acid 97%
(3-Methylbutyl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₁₃BO₂
MW: 115.967 g/mol
MDL Number: MFCD07784365
CAS Number: 98139-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR