(4-(1-(Methoxycarbonyl)cyclopropyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A384912-100MG A384912-250MG
76817-708EA 46.68 USD
76817-708 76817-710
(4-(1-(Methoxycarbonyl)cyclopropyl)phenyl)boronic acid 97%
(4-(1-(Methoxycarbonyl)cyclopropyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BO₄
MW: 220.03 g/mol
MDL Number: MFCD12546570
CAS Number: 1217501-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR