Methyl-2-amino-3-fluorobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-3-fluorobenzoate

A384165-1G A384165-250MG A384165-10G A384165-25G
76817-702EA 22 USD
76817-702 76817-704 77672-904 77884-014
Methyl-2-amino-3-fluorobenzoate 95%
Methyl 2-amino-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.1530 g/mol
MDL Number: MFCD11870179
CAS Number: 144851-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR