1-(tert-Butyl)-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A384158-100MG A384158-250MG A384158-1G
76817-698EA 26.53 USD
76817-698 76817-700 77594-154
1-(tert-Butyl)-1H-pyrazole-3-carboxylic acid 95%
1-(tert-Butyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.19 g/mol
MDL Number: MFCD20261452
CAS Number: 942508-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR