(3-Formyl-2-methoxy-5-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Formyl-2-methoxy-5-methylphenylboronic acid

A384023-1G A384023-250MG A384023-5G
76817-692EA 83 USD
76817-692 76817-694 76817-696
(3-Formyl-2-methoxy-5-methylphenyl)boronic acid 98%
(3-Formyl-2-methoxy-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MDL Number: MFCD08276750
CAS Number: 480424-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR