Methyl-3-methoxy-5-methylbenzoate 98+%
Supplier: Ambeed
|
Warning
|
A383411-1G
A383411-250MG
76817-678EA
21
USD
76817-678
76817-680
Methyl-3-methoxy-5-methylbenzoate 98+%
Methyl 3-methoxy-5-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₃ |
MDL Number:
MFCD09833214 CAS Number: 108593-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...