1-Methylpiperidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A382918-1G A382918-250MG A382918-25G A382918-5G
76817-670EA 44.56 USD
76817-670 76817-672 76817-674 76817-676
1-Methylpiperidine-2-carboxylic acid 95%
1-Methylpiperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD08060082
CAS Number: 7730-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR