2-Methyloxan-4-one 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyldihydro-2H-pyran-4(3H)-one, 2-Methyltetrahydro-4H-pyran-4-one

A380792-1G A380792-250MG
76817-666EA 65.78 USD
76817-666 76817-668
2-Methyloxan-4-one 98%
2-Methyloxan-4-one

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.142 g/mol
MDL Number: MFCD15071428
CAS Number: 1193-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR