2-Bromo-4-fluorothioanisole 97%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-4-fluorophenyl)(methyl)sulphane

A380440-250MG A380440-1G
76817-650EA 35 USD
76817-650 77672-884
2-Bromo-4-fluorothioanisole 97%
2-Bromo-4-fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFS
MDL Number: MFCD09801042
CAS Number: 91524-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR