4-Bromo-5-methylthiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A378787-1G A378787-250MG A378787-5G
76817-626EA 33 USD
76817-626 76817-628 76817-630
4-Bromo-5-methylthiophene-2-carboxylic acid 97%
4-Bromo-5-methylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅BrO₂S
MDL Number: MFCD01859873
CAS Number: 29421-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR