1,4-Dimethoxy-2,3-dimethylbenzene 95%

Supplier: Ambeed
Warning

A377936-10G A377936-1G A377936-250MG
76817-620EA 457 USD
76817-620 76817-622 76817-624
1,4-Dimethoxy-2,3-dimethylbenzene 95%
1,4-Dimethoxy-2,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MDL Number: MFCD00094721
CAS Number: 39021-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR