4-Amino-6-iodo-2-methylpyrimidine 95%

Supplier: Ambeed
Warning

A377577-100MG A377577-1G A377577-250MG
76817-614EA 42.44 USD
76817-614 76817-616 76817-618
4-Amino-6-iodo-2-methylpyrimidine 95%
4-Amino-6-iodo-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₆IN₃
MW: 235.0300 g/mol
MDL Number: MFCD11040178
CAS Number: 943006-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR