3-Benzyl-6-bromo-2-chloroquinoline 97%

Supplier: Ambeed
Warning

A376628-100G A376628-1G A376628-25G A376628-5G
76817-606EA 100 USD
76817-606 76817-608 76817-610 76817-612
3-Benzyl-6-bromo-2-chloroquinoline 97%
3-Benzyl-6-bromo-2-chloroquinoline

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrClN
MDL Number: MFCD22493487
CAS Number: 654655-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR