L-Lysine acetate salt 98%

Supplier: Ambeed
Warning

Synonyms: L-LYSINE, MONOACETATE, (2S)-2,6-diaminohexanoic acid, L-Lysine acetate, L-Lysine Acetate Salt

A375595-1000G A375595-100G A375595-25G A375595-500G
76817-592EA 65.78 USD
76817-592 76817-594 76817-596 76817-598
L-Lysine acetate salt 98%
L-Lysine acetate salt

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₄
MW: 206.242 g/mol
MDL Number: MFCD00039069
CAS Number: 57282-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR