2-Chloroquinolin-8-ol 97%

Supplier: Ambeed
Warning

A375530-100MG A375530-1G A375530-250MG A375530-5G
76817-586EA 20 USD
76817-586 76817-588 76817-590 77884-004
2-Chloroquinolin-8-ol 97%
2-Chloroquinolin-8-ol

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD00168950
CAS Number: 31568-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR