Isoxazole-5-carbonyl chloride 97%

Supplier: Ambeed
Danger

A374137-1G A374137-250MG
76817-574EA 70.02 USD
76817-574 76817-576
Isoxazole-5-carbonyl chloride 97%
Isoxazole-5-carbonyl chloride

Precaución: for Research Use Only

Formula: C₄H₂ClNO₂
MW: 131.52 g/mol
MDL Number: MFCD00067868
CAS Number: 62348-13-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR