(R)-4-Methylpyrrolidin-2-one 98%

Supplier: Ambeed
Warning

A373987-100MG A373987-250MG A373987-1G
76817-564EA 92.3 USD
76817-564 76817-566 77572-116
(R)-4-Methylpyrrolidin-2-one 98%
(R)-4-Methylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₅H₉NO
MW: 99.13 g/mol
MDL Number: MFCD19217255
CAS Number: 260061-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR