N-(tert-Butoxycarbonyl)-D-tyrosine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Tyr-OH, N-Boc-D-tyrosine

A373758-10G A373758-1G A373758-250MG A373758-25G A373758-5G A373758-100G
76817-554EA 49 USD
76817-554 76817-556 76817-558 76817-560 76817-562 77884-001
N-(tert-Butoxycarbonyl)-D-tyrosine 97%
N-(tert-Butoxycarbonyl)-D-tyrosine

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₅
MW: 281.31 g/mol
MDL Number: MFCD00063030
CAS Number: 70642-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR