6-Fluoro-5-iodopyridin-2-ol 95%

Supplier: Ambeed
Warning

A373691-100MG A373691-1G A373691-250MG
76817-548EA 39 USD
76817-548 76817-550 76817-552
6-Fluoro-5-iodopyridin-2-ol 95%
6-Fluoro-5-iodopyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₃FINO
MDL Number: MFCD16611044
CAS Number: 884660-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR