(R)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate sulfate 98%

Supplier: Ambeed
Warning

Synonyms: R-(-)-Clopidogrel Hydrogen sulphate

A373523-250MG A373523-1G A373523-50MG A373523-100MG
76817-544EA 227 USD
76817-544 77613-118 77672-868 77672-866
(R)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate sulfate 98%
(R)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate sulfate

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClNO₆S₂
MDL Number: MFCD17215518
CAS Number: 120202-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR