2,6-Difluoro-3-nitrophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-3-nitrobenzeneboronic acid, (2,6-Difluoro-3-nitrophenyl)boronic acid

A373377-1G A373377-250MG A373377-5G
76817-538EA 41 USD
76817-538 76817-540 76817-542
2,6-Difluoro-3-nitrophenylboronic acid 95%
2,6-Difluoro-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₂NO₄
MDL Number: MFCD12025957
CAS Number: 1150114-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR