2,2'-Bipyridine-6-carbonitrile 98%

Supplier: Ambeed
Warning

A372549-100MG A372549-1G A372549-5G A372549-250MG
76817-530EA 9 USD
76817-530 77613-108 77613-110 77883-998
2,2'-Bipyridine-6-carbonitrile 98%
2,2'-Bipyridine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₁₁H₇N₃
MDL Number: MFCD07364147
CAS Number: 4392-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR