4,5-Difluoro-2-methoxybenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4,5-Difluoro-2-methoxyphenyl)boronic acid, 4,5-Difluoro-2-methoxyphenylboronic acid, 2-Borono-4,5-difluoroanisole

A370168-1G A370168-250MG A370168-25G A370168-5G
76817-504EA 21.99 USD
76817-504 76817-506 76817-508 76817-510
4,5-Difluoro-2-methoxybenzeneboronic acid 98%
4,5-Difluoro-2-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MDL Number: MFCD08275291
CAS Number: 870777-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR