Methyl 2-(2-bromo-4-chlorophenyl)acetate 98%

Supplier: Ambeed
Warning

A368912-100MG A368912-1G A368912-250MG
76817-488EA 23.34 USD
76817-488 76817-490 76817-492
Methyl 2-(2-bromo-4-chlorophenyl)acetate 98%
Methyl 2-(2-bromo-4-chlorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.516 g/mol
MDL Number: MFCD06658159
CAS Number: 115871-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR