2-((4-Bromonaphthalen-1-yl)oxy)acetic acid 95+%

Supplier: Ambeed
Warning

A368603-100MG A368603-1G A368603-250MG
76817-474EA 48.81 USD
76817-474 76817-476 76817-478
2-((4-Bromonaphthalen-1-yl)oxy)acetic acid 95+%
2-((4-Bromonaphthalen-1-yl)oxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₃
MW: 281.10 g/mol
MDL Number: MFCD03129269
CAS Number: 82746-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR