3-(Benzyloxy)-4-methylbenzoic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyloxy-4-methylbenzoic acid

A367073-1G A367073-250MG A367073-5G
76817-458EA 95.49 USD
76817-458 76817-460 76817-462
3-(Benzyloxy)-4-methylbenzoic acid 95+%
3-(Benzyloxy)-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD06656132
CAS Number: 165662-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR