5-Bromo-2-ethoxy-4-pyridineboronic acid 98%

Supplier: Ambeed
Warning

A366022-1G A366022-250MG A366022-5G
76817-434EA 81.99 USD
76817-434 76817-436 76817-438
5-Bromo-2-ethoxy-4-pyridineboronic acid 98%
5-Bromo-2-ethoxy-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BBrNO₃
MDL Number: MFCD07783887
CAS Number: 612845-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR