3-Aminocyclobutanol 96%

Supplier: Ambeed
Danger

A362731-100MG A362731-1G A362731-250MG A362731-5G
76817-428EA 44.56 USD
76817-428 76817-430 76817-432 77613-064
3-Aminocyclobutanol 96%
3-Aminocyclobutanol

Precaución: for Research Use Only

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD09027510
CAS Number: 4640-44-2
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR