4-Benzyloxy-3,5-difluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A361993-1G A361993-250MG A361993-5G A361993-100MG
76817-422EA 33 USD
76817-422 76817-424 76817-426 77883-978
4-Benzyloxy-3,5-difluorophenylboronic acid 97%
4-Benzyloxy-3,5-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
MDL Number: MFCD16295234
CAS Number: 156635-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR