2,4-Dichloro-5,6,7,8-tetrahydroquinazoline-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A359072-100MG A359072-250MG A359072-1G
76817-408EA 119.89 USD
76817-408 76817-410 77672-806
2,4-Dichloro-5,6,7,8-tetrahydroquinazoline-6-carboxylic acid 95%
2,4-Dichloro-5,6,7,8-tetrahydroquinazoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂N₂O₂
MW: 247.07 g/mol
MDL Number: MFCD12198785
CAS Number: 5458-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR