2-((4-Aminopentyl)(ethyl)amino)ethanol 97%

Supplier: Ambeed
Danger

A358348-1G A358348-25G A358348-5G
76817-394EA 6.37 USD
76817-394 76817-396 76817-398
2-((4-Aminopentyl)(ethyl)amino)ethanol 97%
2-((4-Aminopentyl)(ethyl)amino)ethanol

Precaución: for Research Use Only

Formula: C₉H₂₂N₂O
MW: 174.28 g/mol
MDL Number: MFCD03788185
CAS Number: 69559-11-1
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR