3-Bromo-5-fluoro-2-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A358321-1G A358321-5G
76817-392EA 54.11 USD
76817-392 77672-800
3-Bromo-5-fluoro-2-methoxybenzaldehyde 97%
3-Bromo-5-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
MDL Number: MFCD11520672
CAS Number: 1009093-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR