NS-1643 98%

Supplier: Ambeed
Danger

Synonyms: NS1643, 1,3-Bis(2-hydroxy-5-(trifluoromethyl)phenyl)urea

A355887-10MG A355887-25MG A355887-50MG A355887-1MG
76817-368EA 101.85 USD
76817-368 76817-370 76817-372 77613-022
NS-1643 98%
NS-1643

Precaución: for Research Use Only

Formula: C₁₅H₁₀F₆N₂O₃
MW: 380.2 g/mol
MDL Number: MFCD08705418
CAS Number: 448895-37-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR