(R)-1-(3-Fluorophenyl)ethanol 98%

Supplier: Ambeed

A355794-100MG A355794-1G A355794-250MG A355794-5G
76817-360EA 15.92 USD
76817-360 76817-362 76817-364 76817-366
(R)-1-(3-Fluorophenyl)ethanol 98%
(R)-1-(3-Fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.155 g/mol
MDL Number: MFCD03092995
CAS Number: 126534-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR