Isoguvacine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,6-Tetrahydro-4-pyridinecarboxylic acid hydrochloride

A355363-1G A355363-250MG
76817-356EA 113.52 USD
76817-356 76817-358
Isoguvacine hydrochloride 95%
Isoguvacine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉NO₂·HCl
MW: 163.60 g/mol
MDL Number: MFCD00055192
CAS Number: 68547-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR