4-Bromo-6-methylpyridazin-3(2H)-one 97%

Supplier: Ambeed
Warning

A353957-100MG A353957-1G A353957-250MG A353957-5G A353957-10G
76817-350EA 6 USD
76817-350 76817-352 76817-354 77672-776 77883-968
4-Bromo-6-methylpyridazin-3(2H)-one 97%
4-Bromo-6-methylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MDL Number: MFCD09701342
CAS Number: 954240-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR