1-(tert-Butyl)-1H-pyrazole-5-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A352291-100MG
A352291-1G
A352291-250MG
76817-340EA
36.08
USD
76817-340
76817-342
76817-344
1-(tert-Butyl)-1H-pyrazole-5-carbaldehyde 95%
1-(tert-Butyl)-1H-pyrazole-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂O MW: 152.19 g/mol |
MDL Number:
MFCD00103498 CAS Number: 1204355-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...