4-Bromo-6-chloropicolinonitrile 95%

Supplier: Ambeed
Warning

A351710-100MG A351710-1G A351710-250MG A351710-5G
76817-326EA 87 USD
76817-326 76817-328 76817-330 76817-332
4-Bromo-6-chloropicolinonitrile 95%
4-Bromo-6-chloropicolinonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂
MW: 217.45 g/mol
MDL Number: MFCD14582024
CAS Number: 1206247-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR