Ethyl-1-(4-bromophenyl)cyclopropanecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-(4-bromophenyl)cyclopropanecarboxylate

A350191-1G A350191-250MG A350191-5G
76817-316EA 44.56 USD
76817-316 76817-318 76817-320
Ethyl-1-(4-bromophenyl)cyclopropanecarboxylate 95%
Ethyl 1-(4-bromophenyl)cyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrO₂
MW: 269.1300 g/mol
MDL Number: MFCD15143548
CAS Number: 1215205-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR