L-Threonine-tert-butyl ester hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-tert-Butyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OtBu.HCl

A347894-100G A347894-10G A347894-1G A347894-25G A347894-5G
76817-288EA 382 USD
76817-288 76817-290 76817-292 76817-294 76817-296
L-Threonine-tert-butyl ester hydrochloride 95%
L-Threonine-tert-butyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
MDL Number: MFCD00672361
CAS Number: 69320-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR