6-Bromopyrazin-2(1H)-one 98+%

Supplier: Ambeed
Warning

A343654-1G A343654-100MG A343654-250MG
76817-260EA 248 USD
76817-260 77881-943 77881-944
6-Bromopyrazin-2(1H)-one 98+%
6-Bromopyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O
MDL Number: MFCD20921982
CAS Number: 859063-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR