3-Bromo-(5-methoxycarbonyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-(methoxycarbonyl)phenylboronic acid, (3-Bromo-5-(methoxycarbonyl)phenyl)boronic acid

A342578-1G A342578-250MG A342578-5G
76817-256EA 55 USD
76817-256 76817-258 77883-945
3-Bromo-(5-methoxycarbonyl)phenylboronic acid 97%
3-Bromo-(5-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BBrO₄
MW: 258.8617 g/mol
MDL Number: MFCD08689544
CAS Number: 913835-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR