2,4-Dibromo-3-methylpyridine 95%

Supplier: Ambeed
Warning

A339643-1G A339643-250MG
76817-248EA 47 USD
76817-248 76817-250
2,4-Dibromo-3-methylpyridine 95%
2,4-Dibromo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MDL Number: MFCD11044316
CAS Number: 128071-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR