Methyl 2-oxo-1,2-dihydropyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A338249-100MG A338249-250MG
76817-242EA 31.83 USD
76817-242 76817-244
Methyl 2-oxo-1,2-dihydropyrimidine-4-carboxylate 97%
Methyl 2-oxo-1,2-dihydropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.12 g/mol
MDL Number: MFCD21608027
CAS Number: 950514-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR