methyl 5-formylpyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-Formylpyridine-2-carboxylate

A337847-100MG A337847-10G A337847-1G A337847-250MG A337847-25G A337847-5G
76817-230EA 12.74 USD
76817-230 76817-232 76817-234 76817-236 76817-238 76817-240
methyl 5-formylpyridine-2-carboxylate 98%
methyl 5-formylpyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.146 g/mol
MDL Number: MFCD11100669
CAS Number: 55876-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR