Ethyl 2-(N-Methoxy-N-methylamino)-2-oxoacetate 98%

Supplier: Ambeed
Danger

A336983-1G A336983-5G A336983-25G
76817-222EA 21.99 USD
76817-222 76817-224 77883-932
Ethyl 2-(N-Methoxy-N-methylamino)-2-oxoacetate 98%
Ethyl 2-(N-Methoxy-N-methylamino)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄
MDL Number: MFCD11975566
CAS Number: 139507-52-1
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR