(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

A335986-1G A335986-5G
76817-218EA 19.1 USD
76817-218 76817-220
(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00061189
CAS Number: 195447-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR